siRNA Details
| Gene Symbol: C16orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098514.2 | NP_001091984.2 | 1 | 385 | ||
| NM_152459.4 | NP_689672.4 | 1 | 385 | |||
| XM_005255143.3 | XP_005255200.2 | 1 | 211 | |||
| XM_017022972.1 | XP_016878461.1 | 1 | 211 | |||
| XM_017022973.1 | XP_016878462.1 | 1 | 211 | |||
| XM_017022974.1 | XP_016878463.1 | 1 | 211 | |||
| GenBank mRNA | AK075242.1 | - | 1 | 211 | ||
| AK300708.1 | - | 1 | 159 | |||
| AY358483.1 | - | 1 | 350 | |||
| BC033681.1 | - | 1 | 280 | 5190553 | ||

C16orf89
Genomic Map