siRNA Details
| Gene Symbol: EXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286441.1 | NP_001273370.1 | 12 | 3101 | ||
| NM_152596.3 | NP_689809.2 | 10 | 2846 | |||
| XM_006720415.3 | XP_006720478.1 | 11 | 3018 | |||
| XM_011521298.2 | XP_011519600.1 | 13 | 3331 | |||
| XM_011521299.2 | XP_011519601.1 | 13 | 3062 | |||
| XM_011521300.2 | XP_011519602.1 | 10 | 2590 | |||
| XM_011521301.2 | XP_011519603.1 | 12 | 2887 | |||
| XM_011521302.2 | XP_011519604.1 | 7 | 2404 | |||
| XM_017021970.1 | XP_016877459.1 | 10 | 2590 | |||
| XM_017021971.1 | XP_016877460.1 | 10 | 2588 | |||
| GenBank mRNA | AK123414.1 | - | 2 | 2147 | ||
| BC030628.2 | - | 10 | 2786 | 4827307 | ||

EXD1
Genomic Map