siRNA Details
| Gene Symbol: MLKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142497.2 | NP_001135969.1 | 3 | 1184 | ||
| NM_152649.3 | NP_689862.1 | 3 | 1184 | |||
| XM_005255834.3 | XP_005255891.1 | 4 | 1083 | |||
| XM_011522936.2 | XP_011521238.1 | 3 | 1223 | |||
| GenBank mRNA | AK091708.1 | - | 3 | 927 | ||
| AL833362.1 | - | 3 | 601 | |||
| BC028141.1 | - | 3 | 826 | 5214733 | ||

MLKL
Genomic Map