siRNA Details
| Gene Symbol: N4BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153029.3 | NP_694574.3 | 2 | 909 | ||
| XM_011523482.1 | XP_011521784.1 | 2 | 914 | |||
| GenBank mRNA | AB014515.1 | - | 2 | 909 | ||
| BC108288.1 | - | 1 | 470 | |||
| BC152472.1 | - | 2 | 909 | 8862609 | ||

N4BP1
Genomic Map