siRNA Details
| Gene Symbol: LINC00311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038859.1 | 2 | 637 | |||
| GenBank mRNA | BC030801.1 | - | 2 | 637 | 4399117 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00311| Gene Symbol: LINC00311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038859.1 | 2 | 637 | |||
| GenBank mRNA | BC030801.1 | - | 2 | 637 | 4399117 | |