siRNA Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153260.2 | NP_694992.2 | 6 | 1212 | ||
| GenBank mRNA | AK094131.1 | - | 6 | 1206 | ||
| AK094891.1 | - | 5 | 1446 | |||
| BC058935.1 | - | 6 | 1168 | 5578024 | ||
| BX538324.1 | - | 5 | 1358 | |||
| CR749223.1 | - | 6 | 1181 | |||

LRRC57
Genomic Map