siRNA Details
Gene Symbol: FAM71C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153364.3 | NP_699195.1 | 2 | 1667 | ||
GenBank mRNA | BC031221.1 | - | 2 | 1664 | 5272209 | |
DB514813.1 | - | 1 | 171 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM71C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153364.3 | NP_699195.1 | 2 | 1667 | ||
GenBank mRNA | BC031221.1 | - | 2 | 1664 | 5272209 | |
DB514813.1 | - | 1 | 171 |