siRNA Details
| Gene Symbol: LYSMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143917.1 | NP_001137389.1 | 2 | 281 | ||
| NM_153374.2 | NP_699205.1 | 2 | 872 | |||
| XM_017022057.1 | XP_016877546.1 | 2 | 264 | |||
| GenBank mRNA | AK129759.1 | - | 2 | 351 | ||
| AY125955.1 | - | 2 | 861 | |||
| BC022075.1 | - | 2 | 417 | 4799858 | ||
| BC033515.1 | - | 2 | 869 | 5175226 | ||

LYSMD2
Genomic Map