siRNA Details
| Gene Symbol: MINK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024937.3 | NP_001020108.1 | 5 | 630 | ||
| NM_001321236.1 | NP_001308165.1 | 6 | 746 | |||
| NM_015716.4 | NP_056531.1 | 5 | 630 | |||
| NM_153827.4 | NP_722549.2 | 5 | 630 | |||
| NM_170663.4 | NP_733763.1 | 5 | 630 | |||
| XM_006721531.2 | XP_006721594.1 | 5 | 630 | |||
| XM_006721532.2 | XP_006721595.1 | 5 | 630 | |||
| XM_006721536.2 | XP_006721599.1 | 5 | 630 | |||
| XM_011523906.1 | XP_011522208.1 | 5 | 630 | |||
| XM_017024704.1 | XP_016880193.1 | 5 | 630 | |||
| XM_017024705.1 | XP_016880194.1 | 5 | 630 | |||
| XM_017024706.1 | XP_016880195.1 | 5 | 630 | |||
| XM_017024707.1 | XP_016880196.1 | 5 | 431 | |||
| XM_017024708.1 | XP_016880197.1 | 3 | 281 | |||
| GenBank mRNA | AB035698.1 | - | 5 | 386 | ||
| AB041926.1 | - | 5 | 570 | |||
| AB209450.1 | - | 5 | 571 | |||
| AY775058.1 | - | 5 | 386 | |||
| BC034673.1 | - | 5 | 582 | 4384442 | ||
| CA392419.1 | - | 5 | 603 | |||

MINK1
Genomic Map