siRNA Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300814.1 | NP_001287743.1 | 9 | 1509 | ||
| NM_173352.3 | NP_775487.2 | 9 | 1891 | |||
| GenBank mRNA | AK096419.1 | - | 9 | 1891 | ||
| BX647095.1 | - | 10 | 1509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT78| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300814.1 | NP_001287743.1 | 9 | 1509 | ||
| NM_173352.3 | NP_775487.2 | 9 | 1891 | |||
| GenBank mRNA | AK096419.1 | - | 9 | 1891 | ||
| BX647095.1 | - | 10 | 1509 | |||