siRNA Details
| Gene Symbol: DCUN1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173475.3 | NP_775746.1 | 3 | 1738 | ||
| GenBank mRNA | AK123719.1 | - | 3 | 1668 | ||
| BC040442.1 | - | 3 | 1717 | 5590225 | ||
| BX648998.1 | - | 3 | 1619 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCUN1D3| Gene Symbol: DCUN1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173475.3 | NP_775746.1 | 3 | 1738 | ||
| GenBank mRNA | AK123719.1 | - | 3 | 1668 | ||
| BC040442.1 | - | 3 | 1717 | 5590225 | ||
| BX648998.1 | - | 3 | 1619 | |||