siRNA Details
| Gene Symbol: LOC102723968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130763.1 | 6 | 1450 | |||
| GenBank mRNA | AK098560.1 | - | 6 | 1450 | ||
| AK127969.1 | - | 5 | 2464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723968| Gene Symbol: LOC102723968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130763.1 | 6 | 1450 | |||
| GenBank mRNA | AK098560.1 | - | 6 | 1450 | ||
| AK127969.1 | - | 5 | 2464 | |||