siRNA Details
| Gene Symbol: EWSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026949.1 | 1 | 180 | |||
| GenBank mRNA | AK091087.1 | - | 1 | 168 | ||
| BC101594.1 | - | 1 | 231 | |||
| BC101596.1 | - | 1 | 231 | |||
| BX493526.1 | - | 1 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EWSAT1| Gene Symbol: EWSAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026949.1 | 1 | 180 | |||
| GenBank mRNA | AK091087.1 | - | 1 | 168 | ||
| BC101594.1 | - | 1 | 231 | |||
| BC101596.1 | - | 1 | 231 | |||
| BX493526.1 | - | 1 | 181 | |||