siRNA Details
| Gene Symbol: CRYBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004076.4 | NP_004067.1 | 6 | 727 | ||
| XM_011529902.2 | XP_011528204.1 | 6 | 1265 | |||
| XM_017028599.1 | XP_016884088.1 | 5 | 1122 | |||
| GenBank mRNA | BC069479.1 | - | 6 | 697 | 7262237 | |
| BC102021.1 | - | 6 | 684 | 40029651 | ||
| BC102022.1 | - | 6 | 684 | 40029650 | ||
| BC107482.1 | - | 6 | 684 | 40029652 | ||
| BF726163.1 | - | 2 | 235 | |||
| CR456427.1 | - | 5 | 678 | |||

CRYBB3
Genomic Map