siRNA Details
| Gene Symbol: GOLGA8M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282468.1 | NP_001269397.1 | 9 | 716 | ||
| XM_005268284.4 | XP_005268341.1 | 9 | 1515 | |||
| XM_005268285.4 | XP_005268342.1 | 9 | 1515 | |||
| XM_006720654.3 | XP_006720717.1 | 9 | 659 | |||
| XM_011521951.2 | XP_011520253.1 | 10 | 1084 | |||
| XM_011521952.2 | XP_011520254.1 | 9 | 1515 | |||
| XM_011521953.2 | XP_011520255.1 | 9 | 1515 | |||
| XM_011521954.2 | XP_011520256.1 | 9 | 649 | |||
| Gene Symbol: GOLGA8IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024074.1 | 8 | 917 | |||
| GenBank mRNA | AK093104.1 | - | 8 | 917 | ||

GOLGA8M
Genomic Map