siRNA Details
| Gene Symbol: C17orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321399.1 | NP_001308328.1 | 2 | 229 | ||
| NM_173625.4 | NP_775896.3 | 2 | 229 | |||
| XM_011524648.2 | XP_011522950.1 | 2 | 503 | |||
| XM_011524649.2 | XP_011522951.1 | 2 | 503 | |||
| GenBank mRNA | AK096966.1 | - | 2 | 230 | ||
| BC034672.1 | - | 2 | 172 | 5184947 | ||

C17orf78
Genomic Map