siRNA Details
| Gene Symbol: GAS2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303130.1 | NP_001290059.1 | 7 | 689 | ||
| NM_001303131.1 | NP_001290060.1 | 8 | 1176 | |||
| NM_174942.2 | NP_777602.1 | 8 | 810 | |||
| XM_005268804.3 | XP_005268861.1 | 9 | 879 | |||
| XM_011538219.2 | XP_011536521.1 | 8 | 735 | |||
| XM_011538220.2 | XP_011536522.1 | 7 | 2070 | |||
| XM_011538221.2 | XP_011536523.1 | 7 | 920 | |||
| XM_017019206.1 | XP_016874695.1 | 9 | 953 | |||
| GenBank mRNA | AK095594.1 | - | 8 | 1176 | ||
| AK315189.1 | - | 7 | 667 | |||
| BC043366.1 | - | 8 | 760 | 5104644 | ||

GAS2L3
Genomic Map