siRNA Details
| Gene Symbol: KRT73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175068.2 | NP_778238.1 | 4 | 779 | ||
| GenBank mRNA | AJ508776.1 | - | 4 | 779 | ||
| BC109212.2 | - | 4 | 769 | 40007200 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT73| Gene Symbol: KRT73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175068.2 | NP_778238.1 | 4 | 779 | ||
| GenBank mRNA | AJ508776.1 | - | 4 | 779 | ||
| BC109212.2 | - | 4 | 769 | 40007200 | ||