siRNA Details
| Gene Symbol: SERPINA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284275.1 | NP_001271204.1 | 2 | 381 | ||
| NM_175739.3 | NP_783866.2 | 2 | 571 | |||
| XM_011536714.2 | XP_011535016.1 | 2 | 677 | |||
| XM_011536715.2 | XP_011535017.1 | 3 | 541 | |||
| XM_011536716.2 | XP_011535018.1 | 2 | 487 | |||
| XM_011536717.1 | XP_011535019.1 | 2 | 1370 | |||
| GenBank mRNA | AK301082.1 | - | 2 | 465 | ||
| AY185496.1 | - | 2 | 571 | |||
| AY185497.1 | - | 2 | 381 | |||
| AY220118.1 | - | 2 | 485 | |||
| AY220120.1 | - | 2 | 246 | |||
| AY358700.1 | - | 2 | 415 | |||
| BC111498.1 | - | 2 | 474 | 5498091 | ||

SERPINA9
Genomic Map