siRNA Details
| Gene Symbol: FBXL19-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024348.1 | 1 | 2223 | |||
| GenBank mRNA | AK090544.1 | - | 1 | 1137 | ||
| AK092263.1 | - | 1 | 2223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXL19-AS1| Gene Symbol: FBXL19-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024348.1 | 1 | 2223 | |||
| GenBank mRNA | AK090544.1 | - | 1 | 1137 | ||
| AK092263.1 | - | 1 | 2223 | |||