siRNA Details
| Gene Symbol: NLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178844.3 | NP_849172.2 | 4, 5 | 605 | ||
| NR_075083.2 | 4, 5 | 666 | ||||
| XM_017023038.1 | XP_016878527.1 | 4, 5 | 608 | |||
| GenBank mRNA | AK090431.1 | - | 4, 5 | 662 | ||
| AK090476.1 | - | 4, 5 | 605 | |||
| AY601811.1 | - | 4, 5 | 592 | |||
| FJ889357.1 | - | 1, 2 | 181 | |||

NLRC3
Genomic Map