siRNA Details
| Gene Symbol: KRT40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182497.3 | NP_872303.2 | 9 | 1546 | ||
| XM_017024189.1 | XP_016879678.1 | 7 | 3313 | |||
| GenBank mRNA | AJ786658.1 | - | 7 | 1386 | ||
| AK093919.1 | - | 9 | 1720 | |||
| BC150174.1 | - | 9 | 1546 | 40147687 | ||
| BC150191.1 | - | 9 | 1546 | 40148622 | ||

KRT40
Genomic Map