siRNA Details
| Gene Symbol: KRT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268676.3 | XP_005268733.1 | 2 | 635 | ||
| GenBank mRNA | BX537567.1 | - | 2 | 598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT80| Gene Symbol: KRT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268676.3 | XP_005268733.1 | 2 | 635 | ||
| GenBank mRNA | BX537567.1 | - | 2 | 598 | ||