siRNA Details
| Gene Symbol: MYRFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182530.2 | NP_872336.2 | 19, 20 | 2416 | ||
| XM_011539052.2 | XP_011537354.1 | 18, 19 | 2897 | |||
| XM_011539055.1 | XP_011537357.1 | 17, 18 | 1884 | |||
| XM_017018967.1 | XP_016874456.1 | 18, 19 | 2698 | |||
| XM_017018968.1 | XP_016874457.1 | 19, 20 | 3152 | |||
| XM_017018969.1 | XP_016874458.1 | 18, 19 | 2632 | |||
| XM_017018970.1 | XP_016874459.1 | 17, 18 | 2442 | |||
| XM_017018973.1 | XP_016874462.1 | 15, 16 | 2113 | |||
| GenBank mRNA | AK057785.1 | - | 6, 7 | 533 | ||
| BC101820.1 | - | 4, 5 | 368 | |||
| BC143553.1 | - | 4, 5 | 368 | 9052064 | ||

MYRFL
Genomic Map