siRNA Details
| Gene Symbol: MYRFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182530.2 | NP_872336.2 | 21 | 2503 | ||
| XM_011539052.2 | XP_011537354.1 | 20 | 2984 | |||
| XM_011539055.1 | XP_011537357.1 | 19 | 1971 | |||
| XM_017018967.1 | XP_016874456.1 | 20 | 2785 | |||
| XM_017018968.1 | XP_016874457.1 | 21 | 3239 | |||
| XM_017018969.1 | XP_016874458.1 | 20 | 2719 | |||
| XM_017018970.1 | XP_016874459.1 | 19 | 2529 | |||
| XM_017018973.1 | XP_016874462.1 | 17 | 2200 | |||
| GenBank mRNA | AK057785.1 | - | 8 | 620 | ||
| BC101820.1 | - | 6 | 455 | |||
| BC143553.1 | - | 6 | 455 | 9052064 | ||

MYRFL
Genomic Map