siRNA Details
Gene Symbol: C14orf177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182560.2 | NP_872366.2 | 4 | 1074 | ||
GenBank mRNA | AK098639.1 | - | 4 | 1074 | ||
DB523204.1 | - | 1 | 253 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C14orf177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182560.2 | NP_872366.2 | 4 | 1074 | ||
GenBank mRNA | AK098639.1 | - | 4 | 1074 | ||
DB523204.1 | - | 1 | 253 |