siRNA Details
| Gene Symbol: LINC00670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034144.1 | 2 | 329 | |||
| NR_034145.1 | 3 | 491 | ||||
| GenBank mRNA | AK092009.1 | - | 3 | 490 | ||
| BC122562.1 | - | 2 | 316 | |||
| BC122870.1 | - | 2 | 316 | |||
| DB255288.1 | - | 3 | 492 | |||

LINC00670
Genomic Map