siRNA Details
| Gene Symbol: CCDC144B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036647.1 | 3 | 711 | |||
| GenBank mRNA | AK093811.1 | - | 3 | 711 | ||
| BC062767.1 | - | 3 | 705 | |||
| BC101585.1 | - | 3 | 626 | 8069091 | ||
| BC104815.1 | - | 3 | 626 | 8143818 | ||
| BC143561.1 | - | 3 | 626 | 9052072 | ||
| BX647826.1 | - | 3 | 702 | |||
| Gene Symbol: CCDC144A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014695.2 | NP_055510.1 | 3 | 617 | ||
| NR_130142.1 | 3 | 617 | ||||
| XM_011524089.2 | XP_011522391.1 | 3 | 707 | |||
| XM_017025429.1 | XP_016880918.1 | 3 | 634 | |||
| XM_017025430.1 | XP_016880919.1 | 3 | 707 | |||
| GenBank mRNA | BC133019.1 | - | 3 | 617 | 40147041 | |
| BC144224.1 | - | 3 | 617 | 9052744 | ||
| BC144225.1 | - | 3 | 617 | 9052745 | ||
| Gene Symbol: CCDC144CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023380.1 | 3 | 684 | |||
| GenBank mRNA | BC036241.1 | - | 3 | 696 | ||
| Gene Symbol: LOC101929141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025498.1 | XP_016880987.1 | 1 | 169 | ||

CCDC144B
Genomic Map