siRNA Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182614.2 | NP_872420.1 | 4 | 297 | ||
| XM_005272413.3 | XP_005272470.1 | 4 | 323 | |||
| XM_005272415.2 | XP_005272472.1 | 4 | 297 | |||
| XM_017020558.1 | XP_016876047.1 | 4 | 323 | |||
| XM_017020559.1 | XP_016876048.1 | 5 | 1532 | |||
| XM_017020560.1 | XP_016876049.1 | 3 | 260 | |||
| GenBank mRNA | BC018995.2 | - | 4 | 297 | 4300679 | |

TMEM255B
Genomic Map