siRNA Details
| Gene Symbol: LRRC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135217.1 | NP_001128689.1 | 6 | 1065 | ||
| NM_006992.3 | NP_008923.1 | 6 | 893 | |||
| NM_201650.2 | NP_964013.1 | 6 | 893 | |||
| GenBank mRNA | AK223535.1 | - | 6 | 827 | ||
| AK292291.1 | - | 6 | 828 | |||
| AK292367.1 | - | 6 | 893 | |||
| BC029858.1 | - | 6 | 866 | 5170643 | ||
| U72508.1 | - | 6 | 785 | |||

LRRC23
Genomic Map