siRNA Details
| Gene Symbol: PEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000318.2 | NP_000309.1 | 4 | 1632 | ||
| NM_001079867.1 | NP_001073336.1 | 3 | 1600 | |||
| NM_001172086.1 | NP_001165557.1 | 5 | 1638 | |||
| NM_001172087.1 | NP_001165558.1 | 3 | 1496 | |||
| GenBank mRNA | BC000661.2 | - | 4 | 1405 | 3347824 | |
| BC093043.1 | - | 4 | 1418 | 30399827 | ||
| M85038.1 | - | 4 | 1439 | |||
| M86852.1 | - | 4 | 1547 | |||

PEX2
Genomic Map