siRNA Details
| Gene Symbol: ATXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000332.3 | NP_000323.2 | 8 | 1183 | ||
| NM_001128164.1 | NP_001121636.1 | 7 | 1134 | |||
| GenBank mRNA | BC117125.1 | - | 1 | 245 | 40125676 | |
| JX901140.1 | - | 1 | 183 | |||
| X79204.1 | - | 8 | 1147 | |||

ATXN1
Genomic Map