siRNA Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 9 | 3550 | ||
NM_001128164.1 | NP_001121636.1 | 8 | 3501 | |||
GenBank mRNA | X79204.1 | - | 9 | 3517 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 9 | 3550 | ||
NM_001128164.1 | NP_001121636.1 | 8 | 3501 | |||
GenBank mRNA | X79204.1 | - | 9 | 3517 |