siRNA Details
| Gene Symbol: SELE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000450.2 | NP_000441.2 | 14 | 3822 | ||
| GenBank mRNA | BC142677.1 | - | 14 | 3809 | 40148214 | |
| BC142711.1 | - | 14 | 3809 | 40148829 | ||
| M24736.1 | - | 14 | 3782 | |||
| M30640.1 | - | 14 | 3804 | |||

SELE
Genomic Map