siRNA Details
| Gene Symbol: SPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000582.2 | NP_000573.1 | 6 | 1272 | ||
| NM_001040058.1 | NP_001035147.1 | 7 | 1314 | |||
| NM_001040060.1 | NP_001035149.1 | 6 | 1233 | |||
| NM_001251829.1 | NP_001238758.1 | 5 | 1191 | |||
| NM_001251830.1 | NP_001238759.1 | 8 | 1503 | |||
| XM_017008564.1 | XP_016864053.1 | 5 | 1740 | |||
| GenBank mRNA | AF052124.1 | - | 6 | 1194 | ||
| AK057738.1 | - | 7 | 1250 | |||
| AK075463.1 | - | 8 | 1184 | |||
| AK290090.1 | - | 6 | 1169 | |||
| AK295491.1 | - | 8 | 1438 | |||
| AK296035.1 | - | 6 | 1169 | |||
| BC007016.1 | - | 6 | 1208 | 4052438 | ||
| BC017387.1 | - | 7 | 1254 | 3828885 | ||
| BC022844.1 | - | 6 | 1209 | 4284921 | ||
| BC093033.1 | - | 6 | 1225 | 5288027 | ||
| BX648003.1 | - | 6 | 1187 | |||
| J04765.1 | - | 6 | 1215 | |||
| JF412666.1 | - | 7 | 1085 | |||
| JF412667.1 | - | 6 | 1231 | |||
| KR062183.1 | - | 8 | 1438 | |||
| M83248.1 | - | 7 | 1246 | |||
| X13694.1 | - | 7 | 1216 | |||

SPP1
Genomic Map