siRNA Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004161.4 | NP_004152.1 | 4, 5 | 682 | ||
| GenBank mRNA | AF498929.1 | - | 4, 5 | 296 | ||
| AK315495.1 | - | 4, 5 | 491 | |||
| BC000905.2 | - | 4, 5 | 474 | 2900705 | ||
| BT019528.1 | - | 4, 5 | 296 | |||
| BT019529.1 | - | 4, 5 | 296 | |||
| BX571747.1 | - | 4, 5 | 479 | |||
| CR536488.1 | - | 4, 5 | 296 | |||
| M28209.1 | - | 4, 5 | 346 | |||
| X13898.1 | - | 4, 5 | 296 | |||

RAB1A
Genomic Map