siRNA Details
| Gene Symbol: RBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004164.2 | NP_004155.2 | 3 | 330 | ||
| GenBank mRNA | AK291978.1 | - | 3 | 336 | ||
| BC069296.1 | - | 3 | 279 | 7262683 | ||
| BC069361.1 | - | 3 | 279 | 7262695 | ||
| BC069396.1 | - | 3 | 279 | 7262707 | ||
| BC069424.1 | - | 3 | 279 | 7262719 | ||
| BC069447.1 | - | 3 | 279 | 7262731 | ||
| BC069469.1 | - | 3 | 278 | |||
| BC069513.1 | - | 3 | 279 | 7262755 | ||
| BC069522.1 | - | 3 | 279 | 7262672 | ||
| BX282005.1 | - | 5 | 457 | |||
| U13831.1 | - | 3 | 279 | |||

RBP2
Genomic Map