siRNA Details
| Gene Symbol: RIOK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003831.3 | NP_003822.2 | 13 | 2127 | ||
| XM_011526242.1 | XP_011524544.1 | 12 | 1999 | |||
| XM_011526243.1 | XP_011524545.1 | 13 | 1984 | |||
| GenBank mRNA | AF013591.1 | - | 13 | 1603 | ||
| AK057292.1 | - | 13 | 2127 | |||
| AK303938.1 | - | 13 | 1615 | |||
| AK304571.1 | - | 8 | 822 | |||
| AK310783.1 | - | 6 | 752 | |||
| BC028360.1 | - | 12 | 1755 | 4828351 | ||
| BC039729.1 | - | 13 | 1633 | 5745005 | ||
| BX640929.1 | - | 2 | 476 | |||

RIOK3
Genomic Map