siRNA Details
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002787.4 | NP_002778.1 | 8 | 1384 | ||
GenBank mRNA | BC047697.1 | - | 8 | 1338 | 5786518 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002787.4 | NP_002778.1 | 8 | 1384 | ||
GenBank mRNA | BC047697.1 | - | 8 | 1338 | 5786518 |