siRNA Details
| Gene Symbol: PSMD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002814.3 | NP_002805.1 | 5 | 978 | ||
| NM_170750.2 | NP_736606.1 | 5 | 899 | |||
| GenBank mRNA | AB009619.1 | - | 5 | 876 | ||
| AK058108.1 | - | 5 | 952 | |||
| AK295996.1 | - | 5 | 793 | |||
| BC011960.1 | - | 5 | 887 | 3881328 | ||
| D83197.1 | - | 5 | 952 | |||

PSMD10
Genomic Map