siRNA Details
| Gene Symbol: PTPRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161440.2 | NP_001154912.2 | 7 | 1393 | ||
| NM_002842.4 | NP_002833.4 | 9 | 1927 | |||
| XM_011527183.1 | XP_011525485.1 | 9 | 1948 | |||
| XM_011527188.1 | XP_011525490.1 | 8 | 1897 | |||
| XM_011527190.1 | XP_011525492.1 | 9 | 1948 | |||
| XM_017027056.1 | XP_016882545.1 | 8 | 1935 | |||
| XM_017027057.1 | XP_016882546.1 | 8 | 1935 | |||
| XM_017027058.1 | XP_016882547.1 | 8 | 1968 | |||
| XM_017027059.1 | XP_016882548.1 | 8 | 1935 | |||
| XM_017027060.1 | XP_016882549.1 | 8 | 1935 | |||
| XM_017027061.1 | XP_016882550.1 | 6 | 1401 | |||
| XM_017027062.1 | XP_016882551.1 | 8 | 1843 | |||
| XM_017027063.1 | XP_016882552.1 | 8 | 1836 | |||
| XM_017027064.1 | XP_016882553.1 | 8 | 1935 | |||
| GenBank mRNA | AK097257.1 | - | 9 | 1946 | ||
| BC111715.1 | - | 8 | 1343 | 40008777 | ||
| BC111716.1 | - | 9 | 1877 | 40008775 | ||
| D15049.2 | - | 9 | 1901 | |||

PTPRH
Genomic Map