siRNA Details
| Gene Symbol: RAD52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297419.1 | NP_001284348.1 | 11, 12 | 1441 | ||
| NM_001297421.1 | NP_001284350.1 | 9, 10 | 1125 | |||
| NM_134424.3 | NP_602296.2 | 11, 12 | 1329 | |||
| NR_123713.1 | 13, 14 | 1611 | ||||
| XM_005253720.4 | XP_005253777.1 | 12, 13 | 2203 | |||
| XM_005253721.2 | XP_005253778.1 | 11, 12 | 1429 | |||
| XM_011520990.2 | XP_011519292.1 | 12, 13 | 1465 | |||
| XM_011520991.2 | XP_011519293.1 | 11, 12 | 1331 | |||
| XM_011520995.2 | XP_011519297.1 | 7, 8 | 913 | |||
| XM_017019769.1 | XP_016875258.1 | 12, 13 | 1586 | |||
| XM_017019770.1 | XP_016875259.1 | 10, 11 | 1178 | |||
| GenBank mRNA | AF125948.1 | - | 10, 11 | 1501 | ||
| AF125949.1 | - | 11, 12 | 1725 | |||
| AK290047.1 | - | 11, 12 | 1249 | |||
| AK299283.1 | - | 9, 10 | 1072 | |||
| AY540753.1 | - | 10, 11 | 1191 | |||
| BC144458.1 | - | 10, 11 | 1529 | |||
| L33262.1 | - | 11, 12 | 1459 | |||
| U12134.1 | - | 10, 11 | 1222 | |||
| U27516.1 | - | 10, 11 | 1221 | |||

RAD52
Genomic Map