siRNA Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004053.3 | NP_004044.3 | 4 | 950 | ||
| GenBank mRNA | AK095253.1 | - | 1370 | |||
| AK314147.1 | - | 4 | 705 | |||
| BC007340.2 | - | 4 | 686 | 2989807 | ||
| BC050645.1 | - | 4 | 950 | 6011459 | ||
| BC062627.1 | - | 4 | 921 | 5226210 | ||
| L36720.2 | - | 3 | 230 | |||

BYSL
Genomic Map