siRNA Details
| Gene Symbol: CDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317227.1 | NP_001304156.1 | 12 | 3139 | ||
| NM_001317228.1 | NP_001304157.1 | 12 | 3134 | |||
| NM_004061.4 | NP_004052.2 | 15 | 3768 | |||
| XM_011513927.2 | XP_011512229.1 | 13 | 3045 | |||
| XM_017008920.1 | XP_016864409.1 | 14 | 3395 | |||
| XM_017008921.1 | XP_016864410.1 | 15 | 3488 | |||
| GenBank mRNA | BC047608.1 | - | 15 | 3469 | 5751948 | |
| L33477.1 | - | 15 | 3771 | |||

CDH12
Genomic Map