siRNA Details
| Gene Symbol: CXCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002994.4 | NP_002985.1 | 4 | 518 | ||
| GenBank mRNA | BC008376.1 | - | 4 | 469 | 3826939 | |
| BU169770.1 | - | 4 | 489 | |||
| X78686.1 | - | 4 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCL5| Gene Symbol: CXCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002994.4 | NP_002985.1 | 4 | 518 | ||
| GenBank mRNA | BC008376.1 | - | 4 | 469 | 3826939 | |
| BU169770.1 | - | 4 | 489 | |||
| X78686.1 | - | 4 | 476 | |||