siRNA Details
| Gene Symbol: SECTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003004.2 | NP_002995.1 | 5 | 2000 | ||
| XM_005256392.3 | XP_005256449.1 | 5 | 1784 | |||
| XM_011523588.2 | XP_011521890.1 | 5 | 2031 | |||
| GenBank mRNA | AK223068.1 | - | 5 | 1693 | ||
| BC017716.1 | - | 5 | 1694 | 4413387 | ||
| U77643.1 | - | 5 | 1719 | |||

SECTM1
Genomic Map