siRNA Details
| Gene Symbol: SELPLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206609.1 | NP_001193538.1 | 2 | 2118 | ||
| NM_003006.4 | NP_002997.2 | 2 | 2053 | |||
| GenBank mRNA | AK098315.1 | - | 4 | 1972 | ||
| AK127638.1 | - | 3 | 2004 | |||
| AK225644.1 | - | 2 | 1949 | |||
| AK295238.1 | - | 3 | 1835 | |||
| BC029782.1 | - | 3 | 2002 | 5180824 | ||

SELPLG
Genomic Map