siRNA Details
| Gene Symbol: SEMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003007.4 | NP_002998.1 | 2 | 1466 | ||
| GenBank mRNA | J04440.1 | - | 2 | 1382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEMG1| Gene Symbol: SEMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003007.4 | NP_002998.1 | 2 | 1466 | ||
| GenBank mRNA | J04440.1 | - | 2 | 1382 | ||