siRNA Details
| Gene Symbol: SHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003028.2 | NP_003019.2 | 3, 4 | 1630 | ||
| GenBank mRNA | BC136581.1 | - | 3, 4 | 1503 | 9020571 | |
| BC136582.1 | - | 3, 4 | 1503 | 9020572 | ||
| X75342.1 | - | 3, 4 | 1633 | |||

SHB
Genomic Map