siRNA Details
| Gene Symbol: SIAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006610.1 | NP_001006611.1 | 2 | 1175 | ||
| NM_003031.3 | NP_003022.3 | 2 | 803 | |||
| XM_006721246.1 | XP_006721309.1 | 3 | 979 | |||
| XM_011523279.1 | XP_011521581.1 | 6 | 2319 | |||
| GenBank mRNA | AB451253.1 | - | 1 | 686 | ||
| AB451377.1 | - | 1 | 686 | |||
| AK094663.1 | - | 1 | 2140 | |||
| AK225895.1 | - | 2 | 836 | |||
| AK313861.1 | - | 2 | 1189 | |||
| BC035562.1 | - | 2 | 803 | 5224755 | ||
| BC042550.1 | - | 2 | 1145 | 5198176 | ||
| BC044920.1 | - | 6 | 2282 | 5295925 | ||
| BX647064.1 | - | 1 | 1747 | |||
| U63295.1 | - | 1 | 686 | |||
| U76247.1 | - | 2 | 872 | |||

SIAH1
Genomic Map